(S)-(+)-Ethyl-3-piperidinecarboxylate

Supplier: Apollo Scientific
Danger

Synonyms: Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate

OR8970-1G OR8970-5G
APOSOR8970-1GEA 59.1 GBP
APOSOR8970-1G APOSOR8970-5G
(S)-(+)-Ethyl-3-piperidinecarboxylate
(S)-(+)-Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157,21 g/mol
Density: 1,043 g/cm³ (25 °C)
Flash Pt: 76,7 °C
Storage Temperature: Ambient
MDL Number: MFCD00792499
CAS Number: 37675-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR