4-Bromo-3-methylbenzenesulphonyl chloride 97%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Bromo-3-methylbenzene-1-sulphonyl chloride

OR8938-25G OR8938-5G
APOSOR8938-25GEA 243 GBP
APOSOR8938-25G APOSOR8938-5G
4-Bromo-3-methylbenzenesulphonyl chloride 97%
4-Bromo-3-methylbenzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269,55 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD03094696
CAS Number: 72256-93-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR