2-Hydroxychalcone 98%

Supplier: Apollo Scientific
Warning

OR8852-1G OR8852-5G
APOSOR8852-1GEA 19.7 GBP
APOSOR8852-1G APOSOR8852-5G
2-Hydroxychalcone 98%
2-Hydroxychalcone
Formula: C₁₅H₁₂O₂
Melting Pt: 144…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00016449
CAS Number: 644-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR