5-Pyrimidineboronic acid pinacol ester
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Pyrimidine-5-boronic acid pinacol ester, Pyrimidyl-5-boronic acid pinacol ester
OR8851-1G
OR8851-5G
APOSOR8851-1GEA
51.2
GBP
APOSOR8851-1G
APOSOR8851-5G
5-Pyrimidineboronic acid pinacol ester
5-Pyrimidineboronic acid pinacol ester
Formula:
C₁₀H₁₅BN₂O₂ MW: 206,05 g/mol Melting Pt: 60…71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05155223 CAS Number: 321724-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...