Bromopyruvic acid

Supplier: Apollo Scientific
Danger

Synonyms: 3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid

OR8799-10G OR8799-50G
APOSOR8799-10GEA 129 GBP
APOSOR8799-10G APOSOR8799-50G
Bromopyruvic acid
Bromopyruvic acid
Formula: BrH₂CC(O)COOH
MW: 166,96 g/mol
Melting Pt: 75…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002587
CAS Number: 1113-59-3
EINECS: 214-206-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR