4-Methylimidazole 98%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Methyl-1H-imidazole

OR8780-100G OR8780-25G OR8780-500G
APOSOR8780-100GEA 49.9 GBP
APOSOR8780-100G APOSOR8780-25G APOSOR8780-500G
4-Methylimidazole 98%
4-Methylimidazole
Formula: C₄H₆N₂
MW: 82,11 g/mol
Boiling Pt: 263 °C/760mmHg
Melting Pt: 44…47 °C
Density: 1,0416 g/cm³ (25 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00005201
CAS Number: 822-36-6
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR