5-Amino-4,6-dichloropyrimidine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,6-Dichloropyrimidin-5-ylamine
OR8769-100G
OR8769-25G
OR8769-5G
APOSOR8769-100GEA
237
GBP
APOSOR8769-100G
APOSOR8769-25G
APOSOR8769-5G
5-Amino-4,6-dichloropyrimidine
5-Amino-4,6-dichloropyrimidine
Formula:
C₄H₃Cl₂N₃ MW: 163,99 g/mol Melting Pt: 143…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006108 CAS Number: 5413-85-4 EINECS: 226-503-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...