4-(Pyrrolidin-1-ylmethyl)benzaldehyde 97%

Supplier: Apollo Scientific
Danger

OR8676-1G OR8676-250MG
APOSOR8676-1GEA 322 GBP
APOSOR8676-1G APOSOR8676-250MG
4-(Pyrrolidin-1-ylmethyl)benzaldehyde 97%
4-(Pyrrolidin-1-ylmethyl)benzaldehyde
Formula: C₁₂H₁₅NO
MW: 189,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782421
CAS Number: 650628-72-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR