2-Vinylnaphthalene 97%
Supplier: Apollo Scientific
|
Warning
|
OR8644-25G
OR8644-5G
APOSOR8644-25GEA
236
GBP
APOSOR8644-25G
APOSOR8644-5G
2-Vinylnaphthalene 97%
2-Vinylnaphthalene
Formula:
C₁₂H₁₀ Boiling Pt: 128…131 °C/13mmHg Melting Pt: 62…68 °C Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00004125 CAS Number: 827-54-3 EINECS: 212-573-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...