3-Bromo-5-hydroxypyridine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3-pyridinol, 3-Bromo-5-pyridinol, 5-Bromo-pyridin-3-ol

OR8631-25G OR8631-5G OR8631-100G
APOSOR8631-25GEA 72.2 GBP
APOSOR8631-25G APOSOR8631-5G APOSOR8631-100G
3-Bromo-5-hydroxypyridine
3-Bromo-5-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00661305
CAS Number: 74115-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR