5-Bromo-2-(piperidin-1-yl)pyridine

Supplier: Apollo Scientific
Warning

OR8549-1G OR8549-25G OR8549-5G
APOSOR8549-1GEA 46 GBP
APOSOR8549-1G APOSOR8549-25G APOSOR8549-5G
5-Bromo-2-(piperidin-1-yl)pyridine
5-Bromo-2-(piperidin-1-yl)pyridine
Formula: C₁₀H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00234654
CAS Number: 24255-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR