1H-Pyrazole-4-boronic acid 97%

Supplier: Apollo Scientific
Warning

OR8414-1G OR8414-5G OR8414-250MG
APOSOR8414-1GEA 18.5 GBP
APOSOR8414-1G APOSOR8414-5G APOSOR8414-250MG
1H-Pyrazole-4-boronic acid 97%
1H-Pyrazole-4-boronic acid
Formula: C₃H₅BN₂O₂
MW: 111,9000 g/mol
Melting Pt: 146…151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368251
CAS Number: 763120-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR