2-Chloro-4-(hydroxymethyl)thiazole

Supplier: Apollo Scientific
Warning

OR8405-1G OR8405-5G
APOSOR8405-1GEA 269 GBP
APOSOR8405-1G APOSOR8405-5G
2-Chloro-4-(hydroxymethyl)thiazole
2-Chloro-4-(hydroxymethyl)thiazole
Formula: C₄H₄ClNOS
MW: 149,6 g/mol
Boiling Pt: 46 °C/10mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD09265529
CAS Number: 5198-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR