cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Supplier: Apollo Scientific
Synonyms:
cis-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, cis-3-BOC-Aminocyclobutanecarboxylic acid
OR8280-1G
OR8280-250MG
APOSOR8280-1GEA
89.2
GBP
APOSOR8280-1G
APOSOR8280-250MG
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Formula:
C₁₀H₁₇NO₄ MW: 215,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657680 CAS Number: 1008773-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...