2,8-Quinolinediol

Supplier: Apollo Scientific
Warning

Synonyms: 2,8-Dihydroxyquinoline

OR8256-25G OR8256-5G
APOSOR8256-25GEA 72.2 GBP
APOSOR8256-25G APOSOR8256-5G
2,8-Quinolinediol
2,8-Quinolinediol
Formula: C₉H₇NO₂
MW: 161,16 g/mol
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00216696
CAS Number: 15450-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR