Phenylacetylene

Supplier: Apollo Scientific
Danger

OR8217-25G OR8217-100G OR8217-500G
APOSOR8217-25GEA 17.1 GBP
APOSOR8217-25G APOSOR8217-100G APOSOR8217-500G
Phenylacetylene
Phenylacetylene
Formula: C₈H₆
MW: 102,14 g/mol
Boiling Pt: 142…144 °C
Melting Pt: –45 °C
Density: 0,93 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR