4-(Benzyloxy)benzoic acid

Supplier: Apollo Scientific
Warning

OR7984-1G OR7984-25G OR7984-5G
APOSOR7984-1GEA 44.6 GBP
APOSOR7984-1G APOSOR7984-25G APOSOR7984-5G
4-(Benzyloxy)benzoic acid
4-(Benzyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
MW: 228,25 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00016527
CAS Number: 1486-51-7
EINECS: 216-066-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR