4-(4-Pyrimidin-4-yl-1H-pyrazol-1-yl)benzoic acid 95+%
Supplier: Apollo Scientific
OR7884-1G
OR7884-5G
OR7884-25G
APOSOR7884-1GEA
189
GBP
APOSOR7884-1G
APOSOR7884-5G
APOSOR7884-25G
4-(4-Pyrimidin-4-yl-1H-pyrazol-1-yl)benzoic acid 95+%
4-(4-Pyrimidin-4-yl-1H-pyrazol-1-yl)benzoic acid
Formula:
C₁₄H₁₀N₄O₂ Melting Pt: 365…368 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02579383 CAS Number: 849924-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...