Pyridazine-4-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Pyridazinecarboxylic acid
OR7883-1G
OR7883-100MG
OR7883-250MG
APOSOR7883-1GEA
48.6
GBP
APOSOR7883-1G
APOSOR7883-100MG
APOSOR7883-250MG
Pyridazine-4-carboxylic acid
Pyridazine-4-carboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124,10 g/mol Melting Pt: 244 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011576 CAS Number: 50681-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...