2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]quinolizine-6-carboxylic acid 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3-Dihydro-5-oxo-1H,5H-pyrido[3,2,1-ij]quinoline-6-carboxylic acid
OR7868-1G
OR7868-250MG
APOSOR7868-1GEA
178
GBP
APOSOR7868-1G
APOSOR7868-250MG
2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]quinolizine-6-carboxylic acid 95+%
2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]quinolizine-6-carboxylic acid
Formula:
C₁₃H₁₁NO₃ MW: 229,24 g/mol Melting Pt: 255…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD01567810 CAS Number: 386715-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...