4-[(2-Methoxyethyl)amino]-1-methylpiperidine 95+%

Supplier: Apollo Scientific
Warning

OR7785-1G OR7785-5G OR7785-25G
APOSOR7785-1GEA 125 GBP
APOSOR7785-1G APOSOR7785-5G APOSOR7785-25G
4-[(2-Methoxyethyl)amino]-1-methylpiperidine 95+%
4-[(2-Methoxyethyl)amino]-1-methylpiperidine
Formula: C₉H₂₀N₂O
Boiling Pt: 62…64 °C/0,02mmHg
Storage Temperature: Ambient
MDL Number: MFCD01826433
CAS Number: 416887-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR