5-Bromoacenaphthene

Supplier: Apollo Scientific
Warning

Synonyms: 1,2-Dihydro-5-bromoacenaphthylene, 5-Bromo-1,2-dihydroacenaphthylene

OR7751-100G OR7751-25G
APOSOR7751-100GEA 119 GBP
APOSOR7751-100G APOSOR7751-25G
5-Bromoacenaphthene
5-Bromoacenaphthene
Formula: C₁₂H₉Br
MW: 233,11 g/mol
Boiling Pt: 335 °C
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003809
CAS Number: 2051-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR