4-Amino-3-bromobenzonitrile

Supplier: Apollo Scientific
Warning

OR7631-100G OR7631-25G
APOSOR7631-100GEA 124 GBP
APOSOR7631-100G APOSOR7631-25G
4-Amino-3-bromobenzonitrile
4-Amino-3-bromobenzonitrile
Formula: C₇H₅BrN₂
MW: 197,03 g/mol
Melting Pt: 106…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01861386
CAS Number: 50397-74-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR