4-(Methylsulphonyl)benzonitrile 98%

Supplier: Apollo Scientific
Warning

OR7440-1G OR7440-5G OR7440-25G
APOSOR7440-1GEA 13.2 GBP
APOSOR7440-1G APOSOR7440-5G APOSOR7440-25G
4-(Methylsulphonyl)benzonitrile 98%
4-(Methylsulphonyl)benzonitrile
Formula: C₈H₇NO₂S
MW: 181,2100 g/mol
Melting Pt: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00216489
CAS Number: 22821-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR