4-Benzyloxy-3,5-dichlorophenylboronic acid

Supplier: Apollo Scientific
Danger

OR7155-1G OR7155-250MG OR7155-5G
APOSOR7155-1GEA 59.1 GBP
APOSOR7155-1G APOSOR7155-250MG APOSOR7155-5G
4-Benzyloxy-3,5-dichlorophenylboronic acid
4-Benzyloxy-3,5-dichlorophenylboronic acid
Formula: C₁₃H₁₁BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD04039002
CAS Number: 1003298-85-8
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR