2-Amino-1-cyclopentene-1-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Aminocyclopent-1-enecarbonitrile
OR7114-1G
OR7114-25G
OR7114-5G
APOSOR7114-1GEA
46
GBP
APOSOR7114-1G
APOSOR7114-25G
APOSOR7114-5G
2-Amino-1-cyclopentene-1-carbonitrile
2-Amino-1-cyclopentene-1-carbonitrile
Formula:
C₆H₈N₂ MW: 108,144 g/mol Melting Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00517551 CAS Number: 2941-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...