2,4-Diamino-6-hydroxy-5-nitrosopyrimidine 97%

Supplier: Apollo Scientific

OR7055-100G OR7055-25G OR7055-500G OR7055-10G OR7055-50G
APOSOR7055-100GEA 0 GBP
APOSOR7055-100G APOSOR7055-25G APOSOR7055-500G APOSOR7055-10G APOSOR7055-50G
2,4-Diamino-6-hydroxy-5-nitrosopyrimidine 97%
2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
Formula: C₄H₅N₅O₂
MW: 155,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023263
CAS Number: 2387-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR