(+/-)-Dehydrolinalool

Supplier: Apollo Scientific
Warning

Synonyms: 3,7-Dimethyloct-6-en-1-yn-3-ol, (+/-)-3,7-Dimethyl-6-octen-1-yn-3-ol

OR70302-1G OR70302-250MG
APOSOR70302-1GEA 558 GBP
APOSOR70302-1G APOSOR70302-250MG
(+/-)-Dehydrolinalool
(+/-)-Dehydrolinalool
Formula: C₁₀H₁₆O
MW: 152,24 g/mol
Boiling Pt: 197 °C
Density: 0,8787 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00129075
CAS Number: 29171-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR