5-Nitro-o-anisic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methoxy-5-nitrobenzoic acid

OR70137-1G OR70137-5G
APOSOR70137-1GEA 48.6 GBP
APOSOR70137-1G APOSOR70137-5G
5-Nitro-o-anisic acid
5-Nitro-o-anisic acid
Formula: C₈H₇NO₅
MW: 197,15 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00527902
CAS Number: 40751-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR