1-Amino-6-bromoisoquinoline

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromoisoquinolin-1-ylamine

OR70130-250MG OR70130-500MG OR70130-1G
APOSOR70130-250MGEA 112 GBP
APOSOR70130-250MG APOSOR70130-500MG APOSOR70130-1G
1-Amino-6-bromoisoquinoline
1-Amino-6-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374395
CAS Number: 215453-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR