p-Hydrazinobenzenesulphonic acid hemihydrate

Supplier: Apollo Scientific
Warning

Synonyms: Phenylhydrazine-4-sulphonic acid, 4-Hydrazinobenzenesulphonic acid, hemihydrate

OR70100-25G OR70100-100G
APOSOR70100-25GEA 62.9 GBP
APOSOR70100-25G APOSOR70100-100G
p-Hydrazinobenzenesulphonic acid hemihydrate
p-Hydrazinobenzenesulphonic acid hemihydrate
Formula: C₆H₈N₂O₃S
MW: 188,20 g/mol
Melting Pt: 286 °C
Storage Temperature: Ambient
MDL Number: MFCD00480958
CAS Number: 854689-07-9
EINECS: 202-694-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR