Ethyl-4-amino-1-piperidinecarboxylate 96%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-piperidinecarboxylic Acid Ethyl Ester

OR6931-25G OR6931-5G
APOSOR6931-25GEA 269 GBP
APOSOR6931-25G APOSOR6931-5G
Ethyl-4-amino-1-piperidinecarboxylate 96%
Ethyl-4-amino-1-piperidinecarboxylate
Formula: C₈H₁₆N₂O₂
MW: 172,23 g/mol
Boiling Pt: 118…120 °C/6mmHg
Density: 1,004 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006484
CAS Number: 58859-46-4
EINECS: 261-472-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR