Dimethyl(2-piperazin-1-ylethyl)amine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-[2-(Dimethylamino)ethyl]piperazine
OR6907-5G
OR6907-2G
OR6907-10G
OR6907-50G
APOSOR6907-5GEA
0
GBP
APOSOR6907-5G
APOSOR6907-2G
APOSOR6907-10G
APOSOR6907-50G
Dimethyl(2-piperazin-1-ylethyl)amine 98%
Dimethyl(2-piperazin-1-ylethyl)amine
Formula:
C₈H₁₉N₃ MW: 157,26 g/mol Boiling Pt: 90 °C/9mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00167969 CAS Number: 3644-18-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...