1-(2-Phenylethyl)homopiperazine 98%
Supplier: Apollo Scientific
|
Warning
|
OR6881-10G
OR6881-1G
OR6881-5G
OR6881-100MG
OR6881-250MG
OR6881-500MG
APOSOR6881-10GEA
591
GBP
APOSOR6881-10G
APOSOR6881-1G
APOSOR6881-5G
APOSOR6881-100MG
APOSOR6881-250MG
APOSOR6881-500MG
1-(2-Phenylethyl)homopiperazine 98%
1-(2-Phenylethyl)homopiperazine
Formula:
C₁₃H₂₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01320894 CAS Number: 40389-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...