1-(Pyrrolidinocarbonylmethyl)piperazine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Piperazin-1-yl)-1-(pyrrolidin-1-yl)ethan-1-one

OR6878-10G OR6878-50G
APOSOR6878-10GEA 155 GBP
APOSOR6878-10G APOSOR6878-50G
1-(Pyrrolidinocarbonylmethyl)piperazine 97%
1-(Pyrrolidinocarbonylmethyl)piperazine
Formula: C₁₀H₁₉N₃O
Melting Pt: 74…84 °C
Storage Temperature: Ambient
CAS Number: 39890-45-4
EINECS: 254-677-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR