4-(Piperidin-1-ylmethyl)aniline 97%

Supplier: Apollo Scientific
Danger

OR6790-1G OR6790-5G
APOSOR6790-1GEA 60.3 GBP
APOSOR6790-1G APOSOR6790-5G
4-(Piperidin-1-ylmethyl)aniline 97%
4-(Piperidin-1-ylmethyl)aniline
Formula: C₁₂H₁₈N₂
MW: 190,2900 g/mol
Melting Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD03422516
CAS Number: 29608-05-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR