3-Hydroxybenzonitrile

Supplier: Apollo Scientific
Warning

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

OR6704-25G OR6704-5G OR6704-1G
APOSOR6704-25GEA 40.7 GBP
APOSOR6704-25G APOSOR6704-5G APOSOR6704-1G
3-Hydroxybenzonitrile
3-Hydroxybenzonitrile
Formula: C₇H₅NO
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS Number: 873-62-1
EINECS: 212-845-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR