2-Hydroxybenzimidazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Benzimidazolinone, 2(1H)-Benzimidazolone, 2-Hydroxy-1H-benzimidazole
OR6552-5G
OR6552-25G
OR6552-100G
APOSOR6552-5GEA
15.8
GBP
APOSOR6552-5G
APOSOR6552-25G
APOSOR6552-100G
2-Hydroxybenzimidazole 97%
2-Hydroxybenzimidazole
Formula:
C₇H₆N₂O MW: 134,14 g/mol Melting Pt: 314…316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00127894 CAS Number: 615-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...