Pyrrolidin-1-ylacetonitrile

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Pyrrolidin-1-yl)acetonitrile

OR6547-1G OR6547-5G
APOSOR6547-1GEA 19.7 GBP
APOSOR6547-1G APOSOR6547-5G
Pyrrolidin-1-ylacetonitrile
Pyrrolidin-1-ylacetonitrile
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00014098
CAS Number: 29134-29-0
EINECS: 249-463-2
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR