Ethoxyacetic acid 97%
Supplier: Apollo Scientific
Synonyms:
2-Ethoxyacetic acid
OR6531-250G
OR6531-500G
APOSOR6531-250GEA
211
GBP
APOSOR6531-250G
APOSOR6531-500G
Ethoxyacetic acid 97%
Ethoxyacetic acid
Formula:
C₄H₈O₃ MW: 104,11 g/mol Boiling Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004310 CAS Number: 627-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...