N-Butylethylenediamine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-Amino-2-(butylamino)ethane, N-Butyl-1,2-diaminoethane
OR6518-25G
OR6518-100G
APOSOR6518-25GEA
159
GBP
APOSOR6518-25G
APOSOR6518-100G
N-Butylethylenediamine 97%
N-Butylethylenediamine
Formula:
C₆H₁₆N₂ MW: 116,21 g/mol Boiling Pt: 144…146 °C Density: 0,836 g/cm³ (25 °C) Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025602 CAS Number: 19522-69-1 UN: 2734 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...