1-(3-Aminopropyl)piperidine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-(Piperidin-1-yl)propan-1-amine
OR6515-5G
OR6515-1G
APOSOR6515-5GEA
108
GBP
APOSOR6515-5G
APOSOR6515-1G
1-(3-Aminopropyl)piperidine 97%
1-(3-Aminopropyl)piperidine
Formula:
C₈H₁₈N₂ MW: 142,2419 g/mol Boiling Pt: 84…86 °C Density: 0,895 g/cm³ (25 °C) Flash Pt: 82,2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023784 CAS Number: 3529-08-6 EINECS: 222-557-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...