16-Bromopalmitic acid 95%

Supplier: Apollo Scientific
Warning

OR6482-1G OR6482-250MG OR6482-5G
APOSOR6482-1GEA 112 GBP
APOSOR6482-1G APOSOR6482-250MG APOSOR6482-5G
16-Bromopalmitic acid 95%
16-Bromopalmitic acid

Radiopharmaceutical precursor.

Formula: BrH₂C(CH₂)₁₄COOH
MW: 335,32 g/mol
Boiling Pt: 214…217 °C/1mmHg
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00040779
CAS Number: 2536-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR