4-Bromo-1-methyl-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
OR6411-100G
OR6411-25G
APOSOR6411-100GEA
106
GBP
APOSOR6411-100G
APOSOR6411-25G
4-Bromo-1-methyl-1H-pyrazole 97%
4-Bromo-1-methyl-1H-pyrazole
Formula:
C₄H₅BrN₂ MW: 161,00 g/mol Boiling Pt: 185…188 °C Density: 1,558 g/cm³ (25 °C) Flash Pt: 93,3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179565 CAS Number: 15803-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...