Bromoacetic acid 99%

Supplier: Apollo Scientific
Danger

OR6366-250G OR6366-1KG OR6366-2.5KG
APOSOR6366-250GEA 94.4 GBP
APOSOR6366-250G APOSOR6366-1KG APOSOR6366-2.5KG
Bromoacetic acid 99%
Bromoacetic acid
Formula: BrCH₂COOH
MW: 138,95 g/mol
Boiling Pt: 207…209 °C
Melting Pt: 47…49 °C
Density: 1,93 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR