2-Pyrid-4-ylbenzoic acid 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Pyridin-4-ylbenzoic acid
OR6234-1G
OR6234-2.5G
APOSOR6234-1GEA
224
GBP
APOSOR6234-1G
APOSOR6234-2.5G
2-Pyrid-4-ylbenzoic acid 95%
2-Pyrid-4-ylbenzoic acid
Formula:
C₁₂H₉NO₂ Melting Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD08056297 CAS Number: 133362-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...