3-Methyl-1H-pyrazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Formyl-3-methylpyrazole, 3-Methylpyrazole-4-carboxaldehyde
OR6183-10G
OR6183-1G
OR6183-5G
APOSOR6183-10GEA
231
GBP
APOSOR6183-10G
APOSOR6183-1G
APOSOR6183-5G
3-Methyl-1H-pyrazole-4-carbaldehyde 95%
3-Methyl-1H-pyrazole-4-carbaldehyde
Formula:
C5H6N2O MW: 110,12 g/mol Melting Pt: 105…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01313822 CAS Number: 112758-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...