(S)-(-)-3-Acetylthio-2-methylpropionic acid 98%
Supplier: Apollo Scientific
|
Danger
|
OR61506-1G
OR61506-5G
APOSOR61506-1GEA
260
GBP
APOSOR61506-1G
APOSOR61506-5G
(S)-(-)-3-Acetylthio-2-methylpropionic acid 98%
(S)-(-)-3-Acetylthio-2-methylpropionic acid
Formula:
C₆H₁₀O₃S MW: 162,21 g/mol Boiling Pt: 131…132 °C/2,25mmHg Melting Pt: 5 °C Density: 1,178 g/cm³ (25 °C) Flash Pt: >112,8 °C Storage Temperature: Ambient |
CAS Number:
76497-39-7 EINECS: 278-480-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...