4-Chloro-1H-pyrazole 98%

Supplier: Apollo Scientific
Warning

OR61421-100G OR61421-25G
APOSOR61421-100GEA 187 GBP
APOSOR61421-100G APOSOR61421-25G
4-Chloro-1H-pyrazole 98%
4-Chloro-1H-pyrazole
Formula: C₃H₃ClN₂
MW: 102,5200 g/mol
Boiling Pt: 220 °C
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00159634
CAS Number: 15878-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR