5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile

Supplier: Apollo Scientific
Warning

Synonyms: 3-Cyano-5-methyl-2-(2-nitroanilino)thiophene, 5-Methyl-2-((2-nitrophenyl)amino)thiophene-3-carbonitrile

OR61176-25G OR61176-5G
APOSOR61176-25GEA 65.6 GBP
APOSOR61176-25G APOSOR61176-5G
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
Formula: C₁₂H₉N₃O₂S
MW: 259,28 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD06408054
CAS Number: 138564-59-7
EINECS: 421-300-1
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR